Alfa Aesar™ Yttria Stabilized Zirconium Oxide Conical Crucibles, Low Form
Manufacturer: Alfa Aesar 41285-KF
 View all options for this product
Catalog No. AA41285KF
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Diameter (Metric) O.D. (Top): 58mm; O.D. (Base): 41mm
    Height (Metric) 44mm
    Product Type Yttria Stabilized Zirconium Oxide Conical Crucibles, Low Form
    Quantity 1 Piece
    Volume (English) 50mL
  • Description & Specifications
Related Products
Product Content Feedback