Learn More
Alfa Aesar™ Zirconium Oxide Round Bottom Crucibles, Low Form
Manufacturer:  Alfa Aesar 41296KF
 View more versions of this product
Catalog No. AA41296KF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zirconium Oxide Round Bottom Crucibles, Low Form
63mm
200mL
O.D. (Top): 82mm; O.D. (Base): 42mm
1 Piece