Learn More
Selleck Chemical LLCSupplier Diversity Partner Alfuzosin HCl 50mg 81403-68-1  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S140950MG
Alfuzosin HCl is an alpha1 receptor antagonist used to treat benign prostatic hyperplasia (BPH).
Catalog No. 50-631-0