Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfuzosin hydrochloride, Tocris Bioscience™

Catalog Number p-4783727
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
81403-68-1
C19H28ClN5O4
425.914
YTNKWDJILNVLGX-UHFFFAOYSA-N
alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od
71764
CHEBI:32286
N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride
CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl
This item is not returnable. View return policy
81403-68-1
C19H28ClN5O4
425.914
YTNKWDJILNVLGX-UHFFFAOYSA-N
alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od
71764
CHEBI:32286
N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride
CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl

Uroselective α1 antagonist

Functionally uro-selective α1 adrenoceptor antagonist that does not discriminate between α1 subtypes. Inhibits increases in intraurethral pressure caused by phenylephrine-induced contraction by 81% with minor cardiovascular effects. Also relaxes corpus cavernosum tissue (pIC50 = 7.64) in vitro.
Quantity 10 mg
Formula Weight 425.91
Percent Purity >99%
Chemical Name or Material Alfuzosin hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit