Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG13 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548918

Catalog No. PA548918

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
Specifications

Specifications

ALG13
Polyclonal
PBS with 0.09% sodium azide
Q9NP73, Q9D8C3
Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13, Asparagine-linked glycosylation 13 homolog, Glycosyltransferase 28 domain-containing protein 1, UDP-N-acetylglucosamine transferase subunit ALG13 homolog, ALG13, CXorf45, GLT28D1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67574, 79868
Western Blot
Unconjugated
ALG13
Liquid
ALG13
KLH conjugated synthetic peptide between 44-70 amino acids from the Central region of human ALG13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit