Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 208101AP

Catalog No. 50-173-3102


Add to cart

Description

Description

Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
Specifications

Specifications

ALG13
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I0K7, Q9D8C3, Q9NP73
ALG13, CXorf45, GLT28D1, MDS031, YGL047W
Rabbit
IgG
150 μL
-20°C
Primary
300284, 67574, 79868
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ALG13
Liquid
Alg13
ALG13 Fusion Protein Ag14763
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit