Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ALG14 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549657

Catalog No. PA549657

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

May be involved in protein N-glycosylation. May play a role in the second step of the dolichol-linked oligosaccharide pathway. May anchor the catalytic subunit ALG13 to the ER.
Specifications

Specifications

ALG14
Polyclonal
PBS with 0.09% sodium azide
Q96F25
UDP-N-acetylglucosamine transferase subunit ALG14 homolog, ALG14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
199857
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ALG14
Liquid
ALG14
KLH conjugated synthetic peptide between 67-93 amino acids from the Central region of human ALG14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit