Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALG2 Monoclonal Antibody (OTI2A3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525847
Description
ALG2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ALG2 is a member of the glycosyltransferase 1 family. It acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).This gene encodes a member of the glycosyltransferase 1 family. The encoded protein acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).
Specifications
ALG2 | |
Monoclonal | |
0.83 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H553 | |
ALG2 | |
Full length human recombinant protein of ALG2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2A3 | |
Unconjugated | |
ALG2 | |
1110018A23Rik; 1300013N08Rik; Alg2; ALG2 alpha-1,3/1,6-mannosyltransferase; ALG2, alpha-1,3/1,6-mannosyltransferase; ALPG2; alpha-1,3/1,6-mannosyltransferase ALG2; alpha-1,3-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; asparagine-linked glycosylation protein 2 homolog; CDGIi; CMS14; CMSTA3; FLJ14511; FLJ14511 protein; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; hALPG2; homolog of yeast ALG2; im:7145131; LOW QUALITY PROTEIN: alpha-1,3/1,6-mannosyltransferase ALG2; MNCb-5081; NET38; UNQ666/PRO1298 | |
Mouse | |
Affinity Chromatography | |
RUO | |
85365 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction