Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ALG6
Immunogen Synthetic peptides corresponding to ALG6(asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase)) The peptide sequence was selected from the N terminal of ALG6. Peptide sequence PLTAYHSLLCAYVAKFINPDWIALHTSR
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16249520 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16249520UL
20ul Each for $149.00
Add to cart
 
NBP162495 View Documents Novus BiologicalsSupplier Diversity Partner
NBP162495
100 ul Each for $329.00
Add to cart
 
Description

Description

ALG6 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ALG6
Rabbit
Polyclonal
Unconjugated
29929
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to ALG6(asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase)) The peptide sequence was selected from the N terminal of ALG6. Peptide sequence PLTAYHSLLCAYVAKFINPDWIALHTSR
Primary
IgG
alpha-1,3-glucosyltransferase), asparagine-linked glycosylation 6 homolog (S. cerevisiae, asparagine-linked glycosylation 6 homolog (yeast, asparagine-linked glycosylation 6, alpha-1,3-glucosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 6 homolog, CDG1C, dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase, Dolichyl-P-Glc:Man9GlcNAc2-PP-dolichyl glucosyltransferase, dolichyl-P-Glc:Man9GlcNAc2-PP-dolichylglucosyltransferase, EC 2.4.1, EC 2.4.1.-, Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit