Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549039

Catalog No. PA549039

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Adds the second glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation. Transfers glucose from dolichyl phosphate glucose (Dol-P-Glc) onto the lipid-linked oligosaccharide Glc(1)Man(9)GlcNAc(2)-PP-Dol.
Specifications

Specifications

ALG8
Polyclonal
PBS with 0.09% sodium azide
Q9BVK2
Probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1, 3-glucosyltransferase, Asparagine-linked glycosylation protein 8 homolog, Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1, 3-glucosyltransferase, Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase, ALG8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79053
Western Blot
Unconjugated
ALG8
Liquid
ALG8
KLH conjugated synthetic peptide between 15-41 amino acids from the N-terminal region of human ALG8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit