Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALG8 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ALG8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP169028
|
Novus Biologicals
NBP169028 |
100 μL |
Each of 1 for $436.00
|
|
Description
ALG8 Polyclonal specifically detects ALG8 in Rat samples. It is validated for Western Blot.Specifications
ALG8 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
alpha-1,3-glucosyltransferase), asparagine-linked glycosylation 8 homolog (S. cerevisiae, asparagine-linked glycosylation 8 homolog (yeast, asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 8 homolog, CDG1H, dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase, Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase, dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase, EC 2.4.1, EC 2.4.1.-, HUSSY-02, MGC2840, probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase | |
ALG8 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
Q497D1 | |
79053 | |
IgG | |
Affinity Purified | |
59 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title