Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alginate, Mouse anti-Human, ATTO655, Clone: 4B10-1C5, Abnova
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18146
Specifications
Alginate | |
4B10-1C5 | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Liquid | |
Sodium Alginate conjugated to KLH. | |
100 μg | |
Store at -20°C. | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
ATTO655 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Mouse | |
IgG1, kappa | |
RUO | |
Antibody | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title