Learn More
Alginate Mouse anti-Non-species specific, DyLight 350, Clone: 3G4-1F5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259349UV
Catalog No. NBP259349UV
Add to cart
Description
Description
Alginate Monoclonal antibody specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Alginate
3G4-1F5
Western Blot, Immunohistochemistry
Purified
Sodium Alginate conjugated to KLH
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, Immunohistochemistry
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1 Kappa
100 ul
Primary
Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.