Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alginate Antibody (3G4-1F5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259349G
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Alginate | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Western Blot, Immunohistochemistry | |
3G4-1F5 | |
Western Blot, Immunohistochemistry | |
Gum levan, Kelacid, Levan gum, Sazio | |
Sodium Alginate conjugated to KLH | |
100 μL | |
Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction