Learn More
Alginate Mouse anti-Non-species specific, DyLight 650, Clone: 4B10-1C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259348C
Catalog No. NBP259348C
Add to cart
Description
Description
Alginate Monoclonal antibody specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Alginate
4B10-1C5
Western Blot, Immunohistochemistry
Purified
Sodium Alginate conjugated to KLH
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, Immunohistochemistry
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1 Kappa
100 ul
Primary
Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.