Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alginic Acid, NF, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
20 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9005-32-7
(C6H8O7)A(C6H8O7)B
NaN
O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O
9005-32-7
(C6H8O7)A(C6H8O7)B
NaN
O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O

AL172, 9005-32-7, (C6H8O6)n

Spectrum™ Alginic Acid, NF is used as a tablet disintigrant. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 15%
Packaging Fiber Drum
Quantity 25 kg
CAS Max % 100%