Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aliquat™ 336, average MW 442, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
63393-96-4
C25H54ClN
404.164
MFCD00011862
XKBGEWXEAPTVCK-UHFFFAOYSA-M
methyltrioctylammonium chloride,aliquat 336,trioctylmethylammonium chloride,methyl trioctyl ammonium chloride,tomac,tricaprylmethylammonium chloride,capriquat,methyltricaprylylammonium chloride,tricaprylylmethylammonium chloride,trioctyl methyl ammonium chloride
21218
CHEBI:75286
methyl(trioctyl)azanium;chloride
CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-]
63393-96-4
C25H54ClN
404.164
MFCD00011862
XKBGEWXEAPTVCK-UHFFFAOYSA-M
methyltrioctylammonium chloride,aliquat 336,trioctylmethylammonium chloride,methyl trioctyl ammonium chloride,tomac,tricaprylmethylammonium chloride,capriquat,methyltricaprylylammonium chloride,tricaprylylmethylammonium chloride,trioctyl methyl ammonium chloride
21218
CHEBI:75286
methyl(trioctyl)azanium;chloride
CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-]
Phase transfer catalyst
Quantity 1L
Density 0.8800g/mL
Water 5% max.
Flash Point 132°C
Infrared Spectrum Authentic
Color Amber
Refractive Index 1.4655 to 1.4715
Packaging Glass bottle
Fieser 05,460; 06,404; 07,380; 13,192
Formula Weight 442
Physical Form Viscous Liquid
Percent Purity 88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
Chemical Name or Material Aliquat™ 336, Average M.W. 442
Show More Show Less

GHS P Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger