Learn More
Selleck Chemical LLCSupplier Diversity Partner Aliskiren Hemifumarate 50mg 173334-58-2  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S219950MG
Aliskiren hemifumarate is a direct renin inhibitor with IC50 of 1.5 nM.
Catalog No. 50-682-6