Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Complexone 95.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
3952-78-1
C19H15NO8
385.328
MFCD00001202
PWIGYBONXWGOQE-UHFFFAOYSA-N
alizarin complexone,alizarin fluorine blue,alizarin complexon,alizarine complexon,alizarine complexone,alizarinkomplexon,alizarine fluorine blue,3-aminomethylalizarin-n,n-diacetic acid,az-c,alizarin-3-methyliminodiacetic acid
65132
CHEBI:53088
2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O
3952-78-1
C19H15NO8
385.328
MFCD00001202
PWIGYBONXWGOQE-UHFFFAOYSA-N
alizarin complexone,alizarin fluorine blue,alizarin complexon,alizarine complexon,alizarine complexone,alizarinkomplexon,alizarine fluorine blue,3-aminomethylalizarin-n,n-diacetic acid,az-c,alizarin-3-methyliminodiacetic acid
65132
CHEBI:53088
2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O
Color Undesignated
Melting Point 180°C
Quantity 100mg
Formula Weight 385.33
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (HPLC)
Chemical Name or Material Alizarin Complexone

RTECSNumber : AH0585000

TSCA : Yes