Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Red S, 1% (w/v) Aqueous Indicator Solution, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Poly Bottle
130-22-3,7732-18-5
130-22-3,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.