Alizarin Red S sodium salt, Alfa Aesar™

Transition interval: pH 4.6 (yellow) to pH 6.0 (red)

Manufacturer: Alfa Aesar 4204006

Quantity 5g

 View more versions of this product

Catalog No. AA4204006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 130-22-3
    Physical Form Powder
    Molecular Formula C14H7O7SNa
    Formula Weight 342.27
    MDL Number MFCD00013049
    Synonym 3, 4-Dihydroxy-9, 10-dioxo-2-anthracenesulfonic acid sodium salt
    Chemical Name or Material Alizarin Red S sodium salt

    Specifications

    Physical Form Powder
    Formula Weight 342.27
    Quantity 5g
    Color Various
    EINECS Number 204-981-8
    Molecular Formula C14H7O7SNa
    Appearance Various color powder
    MDL Number MFCD00013049
    CAS 130-22-3
    Synonym 3, 4-Dihydroxy-9, 10-dioxo-2-anthracenesulfonic acid sodium salt
    Chemical Name or Material Alizarin Red S sodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit