Learn More
ALK-1 Rat anti-Mouse, Clone: RM0015-1B03, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178138
 View more versions of this product
Catalog No. NBP178138
Add to cart
Description
Description
ALK-1 Monoclonal antibody specifically detects ALK-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
ALK-1
RM0015-1B03
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rat
IgG2
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
NBP1-78138 is reactive to mouse ALK1 extracellular domain.
Mouse
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
LYOPH
Western Blot 1:400-1:800, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100
Purified
ACVRL1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse ALK1 extracellular domain.
Protein G purified
RUO
Primary
94
Reconstitute with 1.0 ml sterilized PBS to a final concentration of 0.2 mg/ml.