Learn More
ALK-1 Rat anti-Mouse, DyLight 405, Clone: RM0015-1B03, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178138V
Catalog No. NBP178138V
Add to cart
Description
Description
ALK-1 Monoclonal antibody specifically detects ALK-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
ALK-1
RM0015-1B03
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ACVRL1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse ALK1 extracellular domain.
Protein G purified
Protein Kinase
Primary
94
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse