Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK-1 Rat anti-Mouse, FITC, Clone: RM0015-1B03, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP178138F

Catalog No. NBP178138F


Add to cart

Description

Description

ALK-1 Monoclonal antibody specifically detects ALK-1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

ALK-1
RM0015-1B03
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
ACVRL1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse ALK1 extracellular domain.
Protein G purified
Protein Kinase
Primary
94
Immunohistochemistry, Immunohistochemistry (Frozen)
FITC
PBS with 0.05% Sodium Azide
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit