Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK-7/Activin Receptor Type 1C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $436.00
Specifications
Antigen | ALK-7/Activin Receptor Type 1C |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
ALK-7/Activin Receptor Type 1C Polyclonal specifically detects ALK-7/Activin Receptor Type 1C in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ALK-7/Activin Receptor Type 1C | |
Western Blot 1:100 - 1:2000 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, ACTR-IC, ACVRLK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
ACVR1C | |
IgG | |
Affinity Purified |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Q8K348.2 | |
130399 | |
Synthetic peptide surrounding amino acid 155 of mouse Activin Receptor Type 1C | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title