Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK/CD246 Mouse anti-Human, Clone: ALK/1503, Novus Biologicals™

Mouse Monoclonal Antibody

$175.29 - $410.59

Specifications

Antigen ALK/CD246
Clone ALK/1503
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25325320
View Documents
Novus Biologicals
NBP25325320UG
20 ug
Each for $175.29
Add to cart
 
NB253253100
View Documents
Novus Biologicals
NBP253253100UG
100 ug
Each for $410.59
Add to cart
 
NB253253200
View Documents
Novus Biologicals
NBP253253200UG
200 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ALK/CD246
Mouse
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
ALK/1503
IgG1 Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit