Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK/CD246 Mouse anti-Human, Clone: ALK/1504, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30777110UG

 View more versions of this product

Catalog No. NB123015


Add to cart

Description

Description

ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

ALK/CD246
ALK/1504
Unconjugated
10 mM PBS with 0.05% BSA
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Recombinant human ALK/CD246 protein fragment (aa200-335)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG2b Kappa
100 μg
Phospho Specific, Protein Kinase, Signal Transduction
Primary
238
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit