Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK/CD246 Rabbit anti-Human, Clone: ALK/3218R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30777310UG
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
ALK/CD246 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
Recombinant human ALK/CD246 protein fragment (aa200-335) | |
100 μg | |
Phospho Specific, Protein Kinase, Signal Transduction | |
238 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
ALK/3218R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title