Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK/CD246 Rabbit anti-Human, Clone: ALK1/2766R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen ALK/CD246
Clone ALK1/2766R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123017
View Documents
Novus Biologicals
NBP30777210UG
100 μg
Each for $406.52
Add to cart
 
NB123018
View Documents
Novus Biologicals
NBP30777220UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

ALK/CD246
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Recombinant full-length human ALK/CD246 protein
Monoclonal
Protein A or G purified
ALK1/2766R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Phospho Specific, Protein Kinase, Signal Transduction
10 mM PBS with 0.05% BSA
238
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit