Learn More
ALK Mouse anti-Human, Clone: OTI3C8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801263
Catalog No. 50-166-5526
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3 ft. half of ALK, derived from chromosome 2, is fused to the 5 ft. portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.
Specifications
Specifications
ALK
Monoclonal
Unconjugated
ALK
Lyophilized
ALK
Human recombit protein fragment corresponding to amino acids 1300-1620 of human ALK produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3C8
PBS with 8% trehalose and no preservative; pH 7.3
Q9UM73
CD246, NBLST3
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
238