Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner Alkaline Phosphatase Activity Fluorometric Assay Kit: High-Throughput Ready Fluorometric Assay to Measure Alkaline Phosphatase (ALP) activity in Cells, Serum, Plasma etc. High sensitivity, Simple & Direct. Detection Limit ~1 μU. 500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K422500
Alkaline phosphatase (ALP) catalyzes the hydrolysis of phosphate esters in alkaline buffer and produces an organic radical and inorganic phosphate. The change in alkaline phosphatase level and activity is associated with a lot of diseases in the liver and bones. Alkaline phosphatase is also a popular enzyme conjugated to secondary antibody in ELISA. In BioVision’s Alkaline Phosphatase Fluorimetric Assay Kit, ALP cleaves the phosphate group of the non-fluorescent 4-Methylumbelliferyl phosphate disodium salt (MUP) substrate resulting in an intense fluorescent signal (Ex/Em = 360nm/440nm). The kit is an ultra sensitive, simple, direct and HTS-ready assay designed to measure ALP activity in serum and bio-samples with detection sensitivity ~1 µU, more sensitive than colorimetric assays. The kit is suitable for both research and drug discovery. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9747