Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline-Iodide-Azide, Pomeroy for Dissolved Oxygen Analysis, Reagents

Catalog No. 01336040
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Catalog No. Quantity
01-336-040 4 L
01-336-041 1 L
01-336-039 500 mL
3 options

Catalog No. 01-336-040

Supplier: Reagents Holdings Llc CA0146004A

Only null left
Add to Cart
Add to Cart

Alkaline-Iodide-Azide, Pomeroy for Dissolved Oxygen Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Colorless
Volume (Metric) 4 L
Packaging Poly Bottle
Quantity 4 L
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.