Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase/ALPP Mouse anti-Human, DyLight™ 550, Clone: HD 8B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP276655R

Catalog No. NBP276655R


Add to cart

Description

Description

Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Alkaline Phosphatase/ALPP
HD 8B6
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
ALPP
Fresh Hep-2 cells followed by booster injections of fresh Hep-2 cells that have had their surface expression of PLAP boosted in culture.
Protein A purified
Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers
Primary
250.0
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes both Regan and Nagao isoenzymes of placental alkaline phosphatase (PLAP).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit