Learn More
Vector LaboratoriesSupplier Diversity Partner Goat Anti-Biotin Alkaline Phosphatase Conjugated 1 ml DFS Item
Manufacturer:  Vector LaboratoriesSupplier Diversity Partner SP3020
This high-affinity polyclonal anti-biotin antibody is produced by hyperimmunizing goats with biotin and is purified by affinity chromatography. The antibody is then cross-linked with alkaline phosphatase (AP) of the highest specific activity using a proprietary conjugation procedure that preserves maximum enzyme activity and antibody specificity. The covalent linkage between the enzyme and antibody is chosen to provide stable, highly active conjugates for immunohistochemical, in situ hybridization, enzyme immunoassay, or transfer blot applications.
Catalog No. NC0079925