Learn More
Abnova™ Alkaline Phosphatase (E. coli)
Used for Enzyme
Manufacturer:  Abnova™ P5305
Catalog No. 89-933-186
Add to cart
Description
Description
#REF!
Specifications
Specifications
Native protein chromatographically purified from E. coli
Liquid
Alkaline Phosphatase (E. coli)
1 mg
RUO
Alkaline Phosphatase Assay
Escherichia coli
Enzyme Activity
In 2.6M ammonium sulfate, pH 8.0
Native protein chromatographically purified from E. coli
Store at 4°C. This product is stable for at least 3 months.
ALKALINE
≥30 units per mg protein (25°C, pH 8.0). One Unit hydrolyzes 1 umole of p-nitrophenol phosphate per minute at 25°C, pH 8.0.
None