Learn More
Molecular Probes™ Alkyne, Succinimidyl Ester (3-Propargyloxypropanoic Acid, Succinimidyl Ester) Non-distribution product as customer accommodation.
alkyne, succinimidyl ester (3-propargyloxypropanoic acid, succinimidyl ester)
Manufacturer:  Molecular Probes™ A10279
Catalog No. A10279
Add to cart
Description
Description
Conjugates prepared with the amine-reactive alkyne, succinimidyl ester can be detected with an azide-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
Specifications
Specifications
Alkyne, Succinimidyl Ester (3-Propargyloxypropanoic Acid, Succinimidyl Ester), 1mg, Yellow, Solid, 225.2 Molecular Weight, Alkyne Dye Type, C10H11NO5 Molecular Formula, Used For Labeling Chemistry
Yellow
Alkyne
≤–20°C
225.2
Solid
Cell analysis, labeling chemistry, labeling amines, amine-reactive fluorophores, click chemistries, cell proliferation with click chemistry
Alkyne, Succinimidyl Ester (3-Propargyloxypropanoic Acid, Succinimidyl Ester)
1mg
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.