Learn More
All-American Hose™ Dura-Pak Attack Hose
For frontline attack duties, C.A.F.S. and foam concentrate applications
Manufacturer:  Snap Tite Hose IncSupplier Diversity Partner DP30X50O25N
 View more versions of this product
Catalog No. 19-819-302
  Check Availability
Add to cart
Specifications
Description
  • Double jacket municipal fire hose
  • Made from 100% high strength, low stretch, air-entangled nylon
  • Has lining of thermoplastic polyurethane for reduced weight while remaining kink resistant
  • Reverse twill weave of inner jacket provides a smooth waterway, comparable only to Ponn Conquest
  • Standard Dura-Cote™ protective treatment increases abrasion, heat, flame and petrochemical resistance
  • Eliminates water pick-up
  • When treated, it is protected from mildew and no drying is required
  • Lining remains flexible to temperatures as low as -40C
  • High resistance to ozone and oxidation
  • Has nylon inner and outer jacket
  • Folds tightly for small spaces, hose beds and hi-rise packs
  • Polyurethane lining material meets National Sanitation Foundation (NSF) Standard 61 for drinking water
Compliance
National Fire Hose Dura-Pak is manufactured in accordance with NFPA 1961 standard, latest edition within our ISO-9001: 2000 certified quality assurance system
Description & Specifications
Specifications
Orange
50 ft.