Learn More
All-American Hose™ N-Dura™ Attack Hose
Suitable for C.A.F.S. and foam solution applications
Manufacturer:  Snap Tite Hose IncSupplier Diversity Partner NP17X50Y15N
 View more versions of this product
Catalog No. 19-819-344
  Check Availability
Add to cart
Specifications
Description
  • Produced from high quality raw materials to exacting standards
  • Double jacket, rubber lined municipal fire hose
  • Made from 100% high strength, low stretch, air-entangled nylon for enhanced abrasion and kink resistance
  • Outer jacket is manufactured with tight weave to eliminate snagging and pulling while on fireground
  • Reverse twill weave of inner jacket provides smooth waterway
  • Delivers low friction loss
  • Standard Dura-Cote™treatment provides heat resistance, reduces moisture absorption, and increases petrochemical resistance
  • Has extruded EPDM rubber tube lining for long term adhesion
  • Lining remains flexible to temperatures as low as -40°C
  • High resistance to ozone and oxidation
Compliance
National Fire Hose N-Dura is manufactured in accordance with NFPA1961 standard, latest edition within our ISO-9001: 2000 certified quality assurance system.
Description & Specifications
Specifications
Yellow
1.75 in.
50 ft.
Attack Hose