Learn More
All-American Hose™ Supreme™ Attack Hose
For use as heavy duty attack hose
Manufacturer:  Snap Tite Hose IncSupplier Diversity Partner FSD30W10025N
 View more versions of this product
Catalog No. 19-819-320
  Check Availability
Add to cart
Specifications
Description
  • Durable, rugged, traditional double jacket fire hose
  • Has double jacket, Ethylene Propylene Diene Monomer (EPDM) rubber lined fire hose
  • Made from 100% high tensile strength polyester yarn
  • Liner is attached to inner jacket using rubber backing that is more flexible and has higher heat resistance
  • Optional Dura-Cote™ treatment increases abrasion, heat, flame and petrochemical resistance
  • When treated, it is protected from mildew and no drying is required
  • Remains flexible to temperatures as low as -40C
  • High resistant to ozone and oxidation
  • Kink resistance exceeds that of lightweight jacketed hose
  • Suitable for use with C.A.F.S. and foam solutions
Certifications
UL listed, FM approval is available on 1.5 in. and 2.5 in. sizes.
Compliance
Ponn Supreme is manufactured in accordance with NFPA 1961 standard, latest edition within our ISO-9001: 2000 certified quality assurance system.
Description & Specifications
Specifications
White (uncoated)
3 in.
100 ft.
Attack Hose