Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DueNorth All-Purpose Industrial Ice/Snow Traction Control

All-Purpose Industrial traction aid offers unparalleled grip, maximum durability, and superior operator safety. Best for those who play hard and work harder in ice and snow.

$42.95 - $50.90

Specifications

Closure Type Pull-on
Color Black
For Use With (Application) Outdoor Surfaces
Material Vulcanized Rubber
Tread Style Pulse Grip
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-060-806
View Documents
DueNorth
V3550370M
Medium
Pair for $42.95
Only null left
Add to Cart
 
19-060-805
View Documents
DueNorth
V3550370L
Large
Pair for $42.95
Only null left
Add to Cart
 
19-060-807
View Documents
DueNorth
V3550370XL
X-Large
Pair for $50.90
Only null left
Add to Cart
 
Description

Description

  • Military grade tungsten carbide replaceable spikes maximize traction and performance
  • Sixteen tungsten carbide spikes per pair—our top-rated replaceable series for tough applications
  • Heel pad features two extra spikes and enlarged heel coverage (versus All Purpose) for increased traction and stability
  • Exclusive specially formulated rubber—provides maximizes flexibility and durability in extreme cold conditions
  • Specially designed pulse grip tread pattern offers added traction characteristics and maximum slip resistance
  • Textured diamond knurling inner lining grips the footwear, keeping traction aids stable and in place
  • Improved heel fit for improved performance
  • Geometrically designed strap webbing and improved shape fit a greater variety of footwear
  • Features molded-in retention strap attachment points
  • For even greater security and fit, Due North offers an optional retention strap (sold separately)
  • Industrial grade sole offers unparalleled grip, maximum durability, and superior operator safety
  • The all-round solution for tough work environment
  • Best for those who play hard and work harder outdoors
Specifications

Specifications

Pull-on
Outdoor Surfaces
Pulse Grip
Unisex
Yes
Black
Vulcanized Rubber
Reusable
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.