Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences All Purpose Reagent Reservoirs, 48mL

Well suited for single and multichannel pipetting.

$39.31 - $121.02

Specifications

Material Polystyrene
Volume (Metric) Well 48 mL
Type Reagent Reservoir
View More Specs

Products 4
Catalog Number Mfr. No. Channels No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Channels No. per Pack Price Quantity & Availability  
03-410-916 View Documents DWK Life Sciences
W845900
8 30 Case of 30 for $39.31
Add to cart
 
03-410-917 View Documents DWK Life Sciences
W845901
8 100 Case of 100 for $121.02
Add to cart
 
03-410-918 View Documents DWK Life Sciences
W845902
12 30 Case of 30 for $39.31
Add to cart
 
03-410-919 View Documents DWK Life Sciences
W845903
12 100 Case of 100 for $121.02
Add to cart
 
Description

Description

  • Made from polystyrene (PE)
  • Available in 8-channel and 12-channel formats
  • Packs of 30 or 100
Specifications

Specifications

Polystyrene
48 mL
Reagent Reservoir
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit