Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

All-rac-a-Tocopheryl Acetate, FCC, 96-102%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
7695-91-2
C31H52O3
472.75
ZAKOWWREFLAJOT-CEFNRUSXSA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
7695-91-2
C31H52O3
472.75
ZAKOWWREFLAJOT-CEFNRUSXSA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

V1048, 7695-91-2, C31H52O3

Spectrum™ All-rac-a-Tocopheryl Acetate, FCC is the synthetic form of Vitamin E. Alpha-tocopheryl acetate is more resistant to oxidation during storage than unesterified tocopherols. The bioavailability of alpha-tocopherol from alpha-tocopheryl acetate is roughly equivalent to that of free alpha-tocopherol. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 100 mL
CAS Max % 1
Percent Purity 96 to 102%