Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

All-rac-alpha-Tocopherol, FCC, 96-102%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
10191-41-0
C29H50O2
430.72
GVJHHUAWPYXKBD-IEOSBIPESA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
10191-41-0
C29H50O2
430.72
GVJHHUAWPYXKBD-IEOSBIPESA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

T1154, 10191-41-0, C29H50O2

Spectrum™ All-rac-alpha-Tocopherol, FCC is the synthetic form of Vitamin E. All-rac-alpha-Tocopherol is often found in fortified foods and nutritional supplements. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 100 mL
CAS Max % 1
Percent Purity 96% to 102%