Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all-trans-Retinal, Spectrum™

Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
100 g
160 g
390 g
Packaging:
Amber Glass Bottle
116-31-4
C20H28O
284.44
NCYCYZXNIZJOKI-OVSJKPMPSA-N
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O
116-31-4
C20H28O
284.44
NCYCYZXNIZJOKI-OVSJKPMPSA-N
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O

R3041, 116-31-4, C20H28O

Spectrum™ all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 160 g
Appearance Yellow powder
CAS Max % 100%