Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all-trans-Retinol, 95%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
500mg
1g
Packaging:
Glass bottle
68-26-8
C20H30O
286.459
MFCD00001552
FPIPGXGPPPQFEQ-OVSJKPMPSA-N
retinol,vitamin a,all-trans-retinol,vitamin a1,alphalin,axerophthol,vitamin a alcohol,oleovitamin a,chocola a,alphasterol
445354
CHEBI:17336
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C
68-26-8
C20H30O
286.459
MFCD00001552
FPIPGXGPPPQFEQ-OVSJKPMPSA-N
retinol,vitamin a,all-trans-retinol,vitamin a1,alphalin,axerophthol,vitamin a alcohol,oleovitamin a,chocola a,alphasterol
445354
CHEBI:17336
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C
Packaging Glass bottle
Color Yellow to Orange
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Beilstein 06, III, 2787
Merck Index 15, 10211
Formula Weight 286.46
Physical Form Powder and Lumps
Percent Purity ≥94%
Chemical Name or Material all-trans-Retinol, 95%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 200-683-7

RTECSNumber : VH6750000

TSCA : TSCA