Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Allantoin, 98%

Click to view available options
Quantity:
250g
1000g
5000g
97-59-6
C4H6N4O3
158.117
MFCD00005260
POJWUDADGALRAB-UHFFFAOYSA-N
allantoin,5-ureidohydantoin,glyoxyldiureide,1-2,5-dioxoimidazolidin-4-yl urea,glyoxyldiureid,allantol,cordianine,sebical,alantan,avc/dienestrolcream
204
CHEBI:15676
(2,5-dioxoimidazolidin-4-yl)urea
C1(C(=O)NC(=O)N1)NC(=O)N
97-59-6
C4H6N4O3
158.117
MFCD00005260
POJWUDADGALRAB-UHFFFAOYSA-N
allantoin,5-ureidohydantoin,glyoxyldiureide,1-2,5-dioxoimidazolidin-4-yl urea,glyoxyldiureid,allantol,cordianine,sebical,alantan,avc/dienestrolcream
204
CHEBI:15676
(2,5-dioxoimidazolidin-4-yl)urea
C1(C(=O)NC(=O)N1)NC(=O)N
Applications
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

Solubility
Slightly soluble in water. Freely soluble in alkalis

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Melting Point ~230°C (decomposition)
Quantity 250g
Beilstein 102364
Merck Index 14,258
Solubility Information Slightly soluble in water. Freely soluble in alkalis
Formula Weight 158.12
Percent Purity 98%
Chemical Name or Material Allantoin

EINECSNumber : 202-592-8

RTECSNumber : YT1600000

TSCA : Yes

Recommended Storage : Ambient temperatures