Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allantoin, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
97-59-6
C4H6N4O3
158.12
POJWUDADGALRAB-UHFFFAOYNA-N
(2,5-dioxoimidazolidin-4-yl)urea
NC(=O)NC1NC(=O)NC1=O
97-59-6
C4H6N4O3
158.12
POJWUDADGALRAB-UHFFFAOYNA-N
(2,5-dioxoimidazolidin-4-yl)urea
NC(=O)NC1NC(=O)NC1=O

A2721, 97-59-6, C4H6N4O3

Spectrum™ Allantoin, USP promotes cell proliferation and wound healing and has a soothing, anti-irritant, and skin protectant effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.1%
Ignition Residue 0.1%
Packaging Poly Pail
Quantity 2.5 kg
CAS Max % 100%
Percent Purity 98.5 to 101%