Learn More
ORS Nasco Allen™ Long-Arm Hex Key Sets
Resist rotational distortion
Supplier: ORS Nasco 02356128
Description
- All hex keys are precision formed to exact tolerances for precise fit
- Alloy steel material
- L-handle, hex tips
- Includes metal box
- Tip size: 1/16 in. (1.5875mm) [min.], 5/64 in. (1.9844mm), 3/32 in. (2.3813mm), 7/64 in. (2.778mm), 1/8 in. (3.175mm), 9/64 in. (3.578mm), 5/32 in. (3.96875mm), 3/16 in. (4.762mm), 7/32 in. (5.556mm), 5/16 in. (7.937mm), 3/8 in. (9.525mm), 1/2 in. (12.7mm) [max.]
Specifications
L-shaped | |
Alloy Steel | |
Black Oxide | |
Metal box |