Learn More
Allergin-1 Mouse anti-Human, Alexa Fluor™ 488, Clone: 767727, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB7638G100UG
Catalog No. FAB7638G10
Add to cart
This item is not returnable. View return policy
Description
Description
Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Allergin-1
767727
Flow Cytometry 0.25-1 μg/106 cells
Q7Z6M3
Allergin1, Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, MCA32-1S2, MILR1, Probable Mast Cell Antigen 32 Homolog
Mouse
IgG1
100 μg
Primary
284021
Human
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant, Conjugated
MILR1
Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227, Accession # Q7Z6M3
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Allergin-1 in direct ELISAs.