Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allergin-1 Mouse anti-Human, Alexa Fluor 700, Clone: 767727, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7638N100UG
This item is not returnable.
View return policy
Description
Allergin-1 Monoclonal specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry.Specifications
Allergin-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q7Z6M3 | |
MILR1 | |
Mouse myeloma cell line NS0-derived recombinant human Allergin-1 Met1-Lys227 Accession # Q7Z6M3 | |
RUO | |
284021 | |
Human | |
IgG1 |
Flow Cytometry | |
767727 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Allergin1, Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, MCA32-1S2, MILR1, Probable Mast Cell Antigen 32 Homolog | |
Mouse | |
100 μg | |
Primary | |
Detects human Allergin-1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)